4-Chlorophenyl phenyl ether
Supplier: Apollo Scientific
|
Danger
|
OR9266-100G
OR9266-25G
OR9266-5G
APOSOR9266-100GEA
227
EUR
APOSOR9266-100G
APOSOR9266-25G
APOSOR9266-5G
4-Chlorophenyl phenyl ether
4-Chlorophenyl phenyl ether
Formula:
C₁₂H₉ClO Boiling Pt: 161…162 °C Density: 1.193 g/cm³ (25 °C) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055431 CAS Number: 7005-72-3 EINECS: 230-281-7 UN: 3082 ADR: 9+EHS,III |