Methyl-3-methyl-1H-pyrazole-4-carboxylate 95%
Supplier: Apollo Scientific
Synonyms: Methyl 3-methyl-1H-pyrazole-4-carboxylate
OR926793-1G OR926793-5G
APOSOR926793-1GEA 125 EUR
APOSOR926793-1G APOSOR926793-5G
Methyl-3-methyl-1H-pyrazole-4-carboxylate 95%
Methyl-3-methyl-1H-pyrazole-4-carboxylate
Formula: C₆H₈N₂O₂
MW: 140.1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07357354
CAS Number: 23170-45-8