2-Ethyl-4,6-dihydroxypyrimidine 95%
Supplier: Apollo Scientific
OR927450-1G
APOSOR927450-1GEA 36.4 EUR
APOSOR927450-1G
2-Ethyl-4,6-dihydroxypyrimidine 95%
2-Ethyl-4,6-dihydroxypyrimidine
Formula: C₆H₈N₂O₂
MW: 140.1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907955
CAS Number: 3709-98-6