[3-(2-Furyl)phenyl]amine hydrochloride 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-(Fur-2-yl)aniline
OR9277-1G OR9277-250MG OR9277-5G
APOSOR9277-1GEA 306 EUR
APOSOR9277-1G APOSOR9277-250MG APOSOR9277-5G
[3-(2-Furyl)phenyl]amine hydrochloride 97%
[3-(2-Furyl)phenyl]amine hydrochloride
Formula: C₁₀H₁₀ClNO
MW: 195.65 g/mol
Melting Pt: 53…54 °C
Storage Temperature: Ambient
MDL Number: MFCD07366478
CAS Number: 102269-42-1
UN: 2811
ADR: 6.1 III