4-(4-Hydroxy-3-methoxyphenyl)-3-buten-2-one 95%
Supplier: Apollo Scientific
Synonyms: Vanillylideneacetone, Dehydrozingerone, 4-(4-Hydroxy-3-methoxyphenyl)but-3-en-2-one
OR927851-25G
APOSOR927851-25GEA 137 EUR
APOSOR927851-25G
4-(4-Hydroxy-3-methoxyphenyl)-3-buten-2-one 95%
4-(4-Hydroxy-3-methoxyphenyl)-3-buten-2-one
Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012210
CAS Number: 1080-12-2