N,N-Dimethyl-1-naphthylamine 95%
Supplier: Apollo Scientific
Synonyms: Dimethyl(1-naphthyl)amine, N,N-Dimethylnaphthalen-1-amine
OR927920-100G OR927920-25G
APOSOR927920-100GEA 469 EUR
APOSOR927920-100G APOSOR927920-25G
N,N-Dimethyl-1-naphthylamine 95%
N,N-Dimethyl-1-naphthylamine
Formula: C₁₂H₁₃N
MW: 171.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003919
CAS Number: 86-56-6