trans-Methyl-4-formylcyclohexanecarboxylate 96%
Supplier: Apollo Scientific
OR928143-10G OR928143-1G OR928143-5G
APOSOR928143-10GEA 676 EUR
APOSOR928143-10G APOSOR928143-1G APOSOR928143-5G
trans-Methyl-4-formylcyclohexanecarboxylate 96%
trans-Methyl-4-formylcyclohexanecarboxylate
Formula: C₉H₁₄O₃
MW: 170.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12965033
CAS Number: 54274-80-5