N-Phenylacrylamide 97%
Supplier: Apollo Scientific
|
Warning
|
OR928266-1G
OR928266-25G
OR928266-5G
APOSOR928266-1GEA
24.8
EUR
APOSOR928266-1G
APOSOR928266-25G
APOSOR928266-5G
N-Phenylacrylamide 97%
N-Phenylacrylamide
Formula:
C₉H₉NO MW: 147.18 g/mol Melting Pt: 103…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00080617 CAS Number: 2210-24-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...