5,6,7,8-Tetrahydroimidazo[1,5-a]pyrazine 96%
Supplier: Apollo Scientific
Warning
OR928290-1G OR928290-250MG
APOSOR928290-1GEA 180 EUR
APOSOR928290-1G APOSOR928290-250MG
5,6,7,8-Tetrahydroimidazo[1,5-a]pyrazine 96%
5,6,7,8-Tetrahydroimidazo[1,5-a]pyrazine
Formula: C₆H₉N₃
MW: 123.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09263955
CAS Number: 297172-19-1