1,8-Naphthyridin-2-amine 98%
Supplier: Apollo Scientific
OR928440-1G OR928440-250MG
APOSOR928440-1GEA 106 EUR
APOSOR928440-1G APOSOR928440-250MG
1,8-Naphthyridin-2-amine 98%
1,8-Naphthyridin-2-amine
Formula: C₈H₇N₃
MW: 145.1613 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01250851
CAS Number: 15992-83-3