Ac-HMBA-linker 98%
Supplier: Apollo Scientific
Synonyms: 4-(Acetoxymethyl)benzoic acid, 4-Acetoxymethylbenzoic acid
OR928683-25G
APOSOR928683-25GEA 459 EUR
APOSOR928683-25G
Ac-HMBA-linker 98%
Ac-HMBA-linker
Formula: C₁₀H₁₀O₄
MW: 194.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01090957
CAS Number: 15561-46-3