3-(2-Nitrophenyl)-1H-pyrazole
Supplier: Apollo Scientific
Warning
OR9289-1G OR9289-250MG
APOSOR9289-1GEA 163 EUR
APOSOR9289-1G APOSOR9289-250MG
3-(2-Nitrophenyl)-1H-pyrazole
3-(2-Nitrophenyl)-1H-pyrazole
Formula: C₉H₇N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00665858
CAS Number: 59844-05-2