[1-(3-Methylpyridin-2-yl)propyl]amine 95%

Supplier: Apollo Scientific

OR929172-1G OR929172-250MG
APOSOR929172-1GEA 719 EUR
APOSOR929172-1G APOSOR929172-250MG
[1-(3-Methylpyridin-2-yl)propyl]amine 95%
[1-(3-Methylpyridin-2-yl)propyl]amine
Formula: C₉H₁₄N₂
Storage Temperature: Ambient
MDL Number: MFCD09971238
CAS Number: 1015846-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR