3,5-Dimethylbenzofuran-2-carbaldehyde 95%
Supplier: Apollo Scientific
OR929341-1G
OR929341-5G
APOSOR929341-1GEA
219
EUR
APOSOR929341-1G
APOSOR929341-5G
3,5-Dimethylbenzofuran-2-carbaldehyde 95%
3,5-Dimethylbenzofuran-2-carbaldehyde
Formula:
C₁₁H₁₀O₂ MW: 174.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07186441 CAS Number: 16817-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...