2-Methyl-4-penten-2-ol 95%
Supplier: Apollo Scientific
Danger
OR929828-5G OR929828-1G
APOSOR929828-5GEA 404 EUR
APOSOR929828-5G APOSOR929828-1G
2-Methyl-4-penten-2-ol 95%
2-Methyl-4-penten-2-ol
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 120 °C
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00021813
CAS Number: 624-97-5
UN: 1987
ADR: 3,III