1-O-Methyl-2-deoxy-D-ribose 95%
Supplier: Apollo Scientific
Warning
OR930083-1G OR930083-25G OR930083-5G
APOSOR930083-1GEA 22 EUR
APOSOR930083-1G APOSOR930083-25G APOSOR930083-5G
1-O-Methyl-2-deoxy-D-ribose 95%
1-O-Methyl-2-deoxy-D-ribose
Formula: C₆H₁₂O₄
Density: 1.224 g/cm³ (25 °C)
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00134161
CAS Number: 60134-26-1