5-(3-Bromophenyl)-3-methyl-1,2,4-oxadiazole 97%
Supplier: Apollo Scientific
Warning
OR9301-1G OR9301-250MG OR9301-5G
APOSOR9301-1GEA 84.1 EUR
APOSOR9301-1G APOSOR9301-250MG APOSOR9301-5G
5-(3-Bromophenyl)-3-methyl-1,2,4-oxadiazole 97%
5-(3-Bromophenyl)-3-methyl-1,2,4-oxadiazole
Formula: C₉H₇BrN₂O
Melting Pt: 95…96 °C
Storage Temperature: Ambient
MDL Number: MFCD08435902
CAS Number: 160377-58-2