2-Chlorothiazole 98%
Supplier: Apollo Scientific
Danger
OR9303-25G OR9303-5G
APOSOR9303-25GEA 59.6 EUR
APOSOR9303-25G APOSOR9303-5G
2-Chlorothiazole 98%
2-Chlorothiazole
Formula: C₃H₂ClNS
MW: 119.57 g/mol
Boiling Pt: 145…146 °C
Density: 1.393 g/cm³ (25 °C)
Flash Pt: 54.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00210701
CAS Number: 3034-52-4
EINECS: 221-228-9
UN: 1993
ADR: 3,III