3-Iodo-1-methyl-1H-pyrazole
Supplier: Apollo Scientific
Danger
OR9305-1G OR9305-5G OR9305-25G
APOSOR9305-1GEA 29 EUR
APOSOR9305-1G APOSOR9305-5G APOSOR9305-25G
3-Iodo-1-methyl-1H-pyrazole
3-Iodo-1-methyl-1H-pyrazole
Formula: C₄H₅IN₂
MW: 208.00 g/mol
Density: 1.936 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD08435890
CAS Number: 92525-10-5