(±)-1,2-Pentanediol 98%
Supplier: Apollo Scientific
Synonyms: (±)-1,2-Dihydroxy pentane, 1,2-Pentanediol
OR930529-25G OR930529-100G OR930529-500G
APOSOR930529-25GEA 21.9 EUR
APOSOR930529-25G APOSOR930529-100G APOSOR930529-500G
(±)-1,2-Pentanediol 98%
(±)-1,2-Pentanediol
Formula: C₅H₁₂O₂
Boiling Pt: 210 °C
Density: 0.97 g/cm³ (25 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00010736
CAS Number: 5343-92-0