H-DL-Phe(3-Cl)-OH 97%
Supplier: Apollo Scientific
Synonyms:
3-Chloro-DL-phenylalanine
OR930842-1G
OR930842-25G
OR930842-5G
APOSOR930842-1GEA
116
EUR
APOSOR930842-1G
APOSOR930842-25G
APOSOR930842-5G
H-DL-Phe(3-Cl)-OH 97%
H-DL-Phe(3-Cl)-OH
Formula:
C₉H₁₀ClNO₂ MW: 199.6 g/mol Storage Temperature: Ambient |
CAS Number:
1956-15-6 |