Boc-Aib-ol 96%
Supplier: Apollo Scientific
Warning
Synonyms: N-Boc-2-amino-2-methyl-1-propanol, N-tert-Butoxycarbonyl-2-amino-2-methyl-1-propanol, tert-Butyl (1-Hydroxy-2-methylpropan-2-yl)carbamate
OR931504-1G OR931504-25G OR931504-5G
APOSOR931504-1GEA 21.9 EUR
APOSOR931504-1G APOSOR931504-25G APOSOR931504-5G
Boc-Aib-ol 96%
Boc-Aib-ol
Formula: C₉H₁₉NO₃
MW: 189.3 g/mol
Melting Pt: 57…62 °C
Storage Temperature: Ambient
MDL Number: MFCD03788641
CAS Number: 102520-97-8