2-Allylisoindoline-1,3-dione 95%
Supplier: Apollo Scientific
Synonyms: N-Allylphthalimide
OR931715-1G OR931715-5G
APOSOR931715-1GEA 101 EUR
APOSOR931715-1G APOSOR931715-5G
2-Allylisoindoline-1,3-dione 95%
2-Allylisoindoline-1,3-dione
Formula: C₁₁H₉NO₂
MW: 187.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00158662
CAS Number: 5428-09-1