Boc-5-chloro-DL-tryptophan 97%
Supplier: Apollo Scientific
OR931877-1G OR931877-250MG OR931877-500MG
APOSOR931877-1GEA 522 EUR
APOSOR931877-1G APOSOR931877-250MG APOSOR931877-500MG
Boc-5-chloro-DL-tryptophan 97%
Boc-5-chloro-DL-tryptophan
Formula: C₁₆H₁₉ClN₂O₄
MW: 338.79 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09264328
CAS Number: 361576-61-6