Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper(I) 95%
Supplier: Apollo Scientific
Warning
Synonyms: Chloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]copper(I), [1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper chloride
OR931916-1G OR931916-5G OR931916-25G
APOSOR931916-1GEA 21.9 EUR
APOSOR931916-1G APOSOR931916-5G APOSOR931916-25G
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper(I) 95%
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper(I)
Formula: C₂₇H₃₆ClCuN₂
MW: 487.60 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD09264276
CAS Number: 578743-87-0