1,2-Diphenylethylamine 97%
Supplier: Apollo Scientific
Synonyms: alpha-Benzylbenzylamine, 1-Amino-1,2-diphenylethane
OR932559-25G OR932559-5G
APOSOR932559-25GEA 877 EUR
APOSOR932559-25G APOSOR932559-5G
1,2-Diphenylethylamine 97%
1,2-Diphenylethylamine
Formula: C₁₄H₁₅N
MW: 197.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008063
CAS Number: 25611-78-3