4-Chloro-2-(1H-pyrazol-3-yl)phenol 95%
Supplier: Apollo Scientific
Synonyms: 3-(5-Chloro-2-hydroxyphenyl)pyrazole
OR932655-5G
APOSOR932655-5GEA 112 EUR
APOSOR932655-5G
4-Chloro-2-(1H-pyrazol-3-yl)phenol 95%
4-Chloro-2-(1H-pyrazol-3-yl)phenol
Formula: C₉H₇ClN₂O
MW: 194.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00209566
CAS Number: 18704-67-1