4-(Trimethylsilyl)-3-butyn-2-one 95%
Supplier: Apollo Scientific
OR932761-25G
APOSOR932761-25GEA 114 EUR
APOSOR932761-25G
4-(Trimethylsilyl)-3-butyn-2-one 95%
4-(Trimethylsilyl)-3-butyn-2-one
Formula: C₇H₁₂OSi
MW: 140.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00190212
CAS Number: 5930-98-3