2-(3-Chloropropyl)-1,3-dioxolane 95%
Supplier: Apollo Scientific
Synonyms: 2-(3-chloropropyl)-1,3-dioxolane, 4-Chlorobutyraldehyde ethylene Acetal
OR932809-1G OR932809-25G OR932809-5G
APOSOR932809-1GEA 139 EUR
APOSOR932809-1G APOSOR932809-25G APOSOR932809-5G
2-(3-Chloropropyl)-1,3-dioxolane 95%
2-(3-Chloropropyl)-1,3-dioxolane
Formula: C₆H₁₁ClO₂
MW: 150.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00043135
CAS Number: 16686-11-6