2-Chloro-N-(1,5-dimethyl-1H-pyrazol-4-yl)acetamide 95%
Supplier: Apollo Scientific
OR933162-1G OR933162-250MG
APOSOR933162-1GEA 502 EUR
APOSOR933162-1G APOSOR933162-250MG
2-Chloro-N-(1,5-dimethyl-1H-pyrazol-4-yl)acetamide 95%
2-Chloro-N-(1,5-dimethyl-1H-pyrazol-4-yl)acetamide
Formula: C₇H₁₀ClN₃O
MW: 187.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04968641
CAS Number: 957261-65-3