(1H-Pyrrol-2-yl)methanamine hydrochloride
Supplier: Apollo Scientific
OR933425-1G
OR933425-250MG
OR933425-5G
APOSOR933425-1GEA
116
EUR
APOSOR933425-1G
APOSOR933425-250MG
APOSOR933425-5G
(1H-Pyrrol-2-yl)methanamine hydrochloride
(1H-Pyrrol-2-yl)methanamine hydrochloride
Formula:
C₅H₉ClN₂ Melting Pt: 120…140 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD20037390 CAS Number: 1351479-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...