3-(4-Nitrophenyl)-1H-pyrazole
Supplier: Apollo Scientific
Warning
OR9335-1G OR9335-250MG
APOSOR9335-1GEA 65.4 EUR
APOSOR9335-1G APOSOR9335-250MG
3-(4-Nitrophenyl)-1H-pyrazole
3-(4-Nitrophenyl)-1H-pyrazole
Formula: C₉H₇N₃O₂
Melting Pt: 194…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00665859
CAS Number: 20583-31-7