3-Chloro-o-anisidine 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Chloro-2-methoxyaniline
OR9341-1G OR9341-25G OR9341-5G
APOSOR9341-1GEA 21.9 EUR
APOSOR9341-1G APOSOR9341-25G APOSOR9341-5G
3-Chloro-o-anisidine 95+%
3-Chloro-o-anisidine
Formula: C₇H₈ClNO
MW: 157.6 g/mol
Boiling Pt: 246 °C
Density: 1.231 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00060607
CAS Number: 51114-68-2