2-Amino-9,10-anthraquinone 80%
Supplier: Apollo Scientific
Synonyms: 2-Aminoanthraquinone
OR936416-25G OR936416-500G OR936416-1G OR936416-5G
APOSOR936416-25GEA 229 EUR
APOSOR936416-25G APOSOR936416-500G APOSOR936416-1G APOSOR936416-5G
2-Amino-9,10-anthraquinone 80%
2-Amino-9,10-anthraquinone
Formula: C₁₄H₉NO₂
MW: 223.23 g/mol
Boiling Pt: 473
Melting Pt: 292…302 °C
Flash Pt: 283 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001233
CAS Number: 117-79-3