S-Ethyl thioacetate 98%
Supplier: Apollo Scientific
OR936522-100G OR936522-25G
APOSOR936522-100GEA 290 EUR
APOSOR936522-100G APOSOR936522-25G
S-Ethyl thioacetate 98%
S-Ethyl thioacetate
Formula: C₄H₈OS
MW: 104.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00015178
CAS Number: 625-60-5