3-Mercapto-4-methyl-4H-1,2,4-triazole 95%

Supplier: Apollo Scientific

Synonyms: 4-Methyl-4H-1,2,4-triazole-3-thiol

OR936548-100G OR936548-25G
APOSOR936548-100GEA 502 EUR
APOSOR936548-100G APOSOR936548-25G
3-Mercapto-4-methyl-4H-1,2,4-triazole 95%
3-Mercapto-4-methyl-4H-1,2,4-triazole
Formula: C₃H₅N₃S
MW: 115.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010069
CAS Number: 24854-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR