4-Benzoyloxy-2-azetidinone 95%

Supplier: Apollo Scientific

OR936652-10G OR936652-1G OR936652-5G
APOSOR936652-10GEA 812 EUR
APOSOR936652-10G APOSOR936652-1G APOSOR936652-5G
4-Benzoyloxy-2-azetidinone 95%
4-Benzoyloxy-2-azetidinone
Formula: C₁₀H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD00012317
CAS Number: 28562-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR