2,2,8,8-Tetramethyl-3,6-nonadiyn-5-ol 95%
Supplier: Apollo Scientific
Synonyms: 1,3-Dineopentylidyne-2-propanol, 1,5-Di-tert-butyl-1,4-pentadiyn-3-ol
OR936687-1G OR936687-5G
APOSOR936687-1GEA 270 EUR
APOSOR936687-1G APOSOR936687-5G
2,2,8,8-Tetramethyl-3,6-nonadiyn-5-ol 95%
2,2,8,8-Tetramethyl-3,6-nonadiyn-5-ol
Formula: C₁₃H₂₀O
MW: 192.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797185
CAS Number: 50428-39-2