Benzenesulphonamide 98%
Supplier: Apollo Scientific
Warning
OR936811-100G OR936811-250G
APOSOR936811-100GEA 21.9 EUR
APOSOR936811-100G APOSOR936811-250G
Benzenesulphonamide 98%
Benzenesulphonamide
Formula: C₆H₇NO₂S
MW: 157.19 g/mol
Melting Pt: 149…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00007930
CAS Number: 98-10-2