trans-1-Methoxy-3-trimethylsilyloxy-1,3-butadiene 95%
Supplier: Apollo Scientific
OR936898-5G
APOSOR936898-5GEA 250 EUR
APOSOR936898-5G
trans-1-Methoxy-3-trimethylsilyloxy-1,3-butadiene 95%
trans-1-Methoxy-3-trimethylsilyloxy-1,3-butadiene
Formula: C₈H₁₆O₂Si
MW: 172.299 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008498
CAS Number: 54125-02-9