Ethyl-2-(chloromethyl)acrylate 95%
Supplier: Apollo Scientific
Synonyms: 2-(Chloromethyl)acrylic Acid Ethyl Ester
OR936949-1G OR936949-250MG
APOSOR936949-1GEA 567 EUR
APOSOR936949-1G APOSOR936949-250MG
Ethyl-2-(chloromethyl)acrylate 95%
Ethyl-2-(chloromethyl)acrylate
Formula: C₆H₉ClO₂
MW: 148.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01321155
CAS Number: 17435-77-7