Butyl vinyl ether 98%
Supplier: Apollo Scientific
OR936968-25G
OR936968-5G
APOSOR936968-25GEA
87
EUR
APOSOR936968-25G
APOSOR936968-5G
Butyl vinyl ether 98%
Butyl vinyl ether
Formula:
C₆H₁₂O Boiling Pt: 94 °C Melting Pt: –92 °C Density: 0.774 g/cm³ (25 °C) Flash Pt: –12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009454 CAS Number: 111-34-2 |