1-sec-Butyl-2-nitrobenzene 95%
Supplier: Apollo Scientific
OR937123-10G OR937123-1G OR937123-5G
APOSOR937123-10GEA 877 EUR
APOSOR937123-10G APOSOR937123-1G APOSOR937123-5G
1-sec-Butyl-2-nitrobenzene 95%
1-sec-Butyl-2-nitrobenzene
Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Storage Temperature: Ambient
CAS Number: 19370-34-4