4-Nitrobenzylamine hydrochloride 97%
Supplier: Apollo Scientific
Warning
Synonyms: (4-Nitrophenyl)methanamine hydrochloride
OR937388-1G OR937388-5G
APOSOR937388-1GEA 21.9 EUR
APOSOR937388-1G APOSOR937388-5G
4-Nitrobenzylamine hydrochloride 97%
4-Nitrobenzylamine hydrochloride
Formula: C₇H₉ClN₂O₂
MW: 188.62 g/mol
Melting Pt: 265 °C
Storage Temperature: Ambient
MDL Number: MFCD00012863
CAS Number: 18600-42-5