3,6-Dimethyl-1-heptyn-3-ol 95%
Supplier: Apollo Scientific
Synonyms: (1,4-Dimethyl-1-hydroxypentyl)acetylene, 3,6-Dimethylhept-1-yn-3-ol
OR937500-1G OR937500-5G
APOSOR937500-1GEA 82.8 EUR
APOSOR937500-1G APOSOR937500-5G
3,6-Dimethyl-1-heptyn-3-ol 95%
3,6-Dimethyl-1-heptyn-3-ol
Formula: C₉H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD00041608
CAS Number: 19549-98-5