Tetrahydrofurfuryl propionate 97%
Supplier: Apollo Scientific
Synonyms: (Tetrahydrofuran-2-yl)methyl propionate
OR937571-100G OR937571-25G OR937571-500G
APOSOR937571-100GEA 97.3 EUR
APOSOR937571-100G APOSOR937571-25G APOSOR937571-500G
Tetrahydrofurfuryl propionate 97%
Tetrahydrofurfuryl propionate
Formula: C₈H₁₄O₃
MW: 158.1950 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047080
CAS Number: 637-65-0