Butylsuccinic anhydride 95%
Supplier: Apollo Scientific
OR937809-10G
OR937809-1G
OR937809-5G
APOSOR937809-10GEA
864
EUR
APOSOR937809-10G
APOSOR937809-1G
APOSOR937809-5G
Butylsuccinic anhydride 95%
Butylsuccinic anhydride
Formula:
C₈H₁₂O₃ Storage Temperature: Ambient |
CAS Number:
2035-76-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...