Potassium isopropylxanthate 95%

Supplier: Apollo Scientific

Synonyms: Isopropylxanthic Acid Potassium Salt

OR938096-25G OR938096-5G
APOSOR938096-25GEA 98.6 EUR
APOSOR938096-25G APOSOR938096-5G
Potassium isopropylxanthate 95%
Potassium isopropylxanthate
Formula: C₄H₈OS₂
Storage Temperature: Ambient
MDL Number: MFCD00004930
CAS Number: 140-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR