4-Chloro-2,1,3-benzoxadiazole 95%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Chlorobenzofurazan
OR938160-10G OR938160-1G OR938160-5G
APOSOR938160-10GEA 434 EUR
APOSOR938160-10G APOSOR938160-1G APOSOR938160-5G
4-Chloro-2,1,3-benzoxadiazole 95%
4-Chloro-2,1,3-benzoxadiazole
Formula: C₆H₃ClN₂O
MW: 154.55 g/mol
Melting Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00191329
CAS Number: 7116-16-7