2-(2-Bromoethyl)-2-methyl-1,3-dioxolane 96%
Supplier: Apollo Scientific
Warning
OR938175-1G OR938175-25G OR938175-5G
APOSOR938175-1GEA 69.6 EUR
APOSOR938175-1G APOSOR938175-25G APOSOR938175-5G
2-(2-Bromoethyl)-2-methyl-1,3-dioxolane 96%
2-(2-Bromoethyl)-2-methyl-1,3-dioxolane
Formula: C₆H₁₁BrO₂
MW: 195.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00191321
CAS Number: 37865-96-6